Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mystaire™ Room Air Purifier
SDP

Catalog No. p-4707061 Shop All Mystaire Products
Click to view available options
Type:
Dual-Replacement Capture Arm
Fume Extractor
Single-Replacement Capture Arm

Target fumes at the source or eliminate room air quality issues.

  • Sparkless and brushless centrifugal fan for quiet operation
  • Epoxy coated aluminum construction
  • HEPA and/or Carbon Filtration
  • HEPA or carbon filter depending on application
  • Lightweight and portable
  • Fume Extractor: One or two adjustable capture arms for easy movement to the source of contamination

Specifications

For Use With (Equipment) Mystaire Ductless Workstation
Type Fume Extractor
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.