Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rootletin Chicken anti-Mouse, Human, Porcine, Polyclonal, MilliporeSigma™

Catalog No. ABN1686MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1686MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1686MI Supplier MilliporeSigma Supplier No. ABN1686
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Anti-Rootletin Antibody is a highly specific chicken polyclonal antibody that targets Rootletin and has been tested in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rootletin
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Crocc,Kiaa0445
Host Species Chicken
Immunogen His-tagged recombinant fragment corresponding to 188 amino acids from the C-terminal region of murine Rootletin.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_742120.2
Test Specificity This chicken polyclonal antibody detects Rootletin in mouse retina. It targets an epitope within 188 amino acids from the C-terminal region.
Target Species Mouse, Human, Pig
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.