Learn More
Description
Specifications
Specifications
| Antigen | Rootletin |
| Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Gene Symbols | Crocc,Kiaa0445 |
| Host Species | Rabbit |
| Immunogen | His-tagged recombinant fragment corresponding to 188 amino acids from the C-terminal region of murine Rootletin. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.