Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROPN1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562632
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562632 Supplier Invitrogen™ Supplier No. PA562632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDLIQWGADY FEALSRGETP PVRERSERVA LCNWAELTPE LLKIL Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 78%.

ROPN1B belongs to the ropporin family and contains 1 RIIa domain. ROPN1B interacts with RHPN1 and AKAP3. It may interact with SPA17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROPN1B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ROPN1B
Gene Accession No. Q9BZX4
Gene Alias AKAP-binding sperm protein ropporin; rhophilin associated tail protein 1B; rhophilin-associated protein 1B; ROPN1B; ropporin, rhophilin associated protein 1B; ropporin-1B; testis tissue sperm-binding protein Li 83P
Gene Symbols ROPN1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ROPN1B. Recombinant protein control fragment (Product #RP-99035).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152015
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.