Learn More
Invitrogen™ ROPN1B Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 83%.
Specifications
Specifications
| Antigen | ROPN1B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | ROPN1B |
| Gene Accession No. | Q9BZX4 |
| Gene Alias | AKAP-binding sperm protein ropporin; rhophilin associated tail protein 1B; rhophilin-associated protein 1B; ROPN1B; ropporin, rhophilin associated protein 1B; ropporin-1B; testis tissue sperm-binding protein Li 83P |
| Gene Symbols | ROPN1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.