Learn More
Invitrogen™ ROPN1L Polyclonal Antibody

Description
Immunogen sequence: PDTMFCAQQI HIPPELPDIL KQFTKAAIRT QPADVLRWSA GYFSALSRGD PLPVKDRMEM PTATQKTDTG LTQGLL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Specifications
Specifications
| Antigen | ROPN1L |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Ropn1l |
| Gene Accession No. | Q96C74 |
| Gene Alias | Akapasp; AKAP-associated sperm protein; A-kinase anchoring protein-associated sperm protein; ASP; AV047578; radial spoke head 11 homolog; rhophilin associated tail protein 1 like; rhophilin associated tail protein 1-like; ROPN1L; ROPN1-like protein; ropporin 1-like; ropporin-1-like protein; RSPH11 |
| Gene Symbols | Ropn1l |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.