Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Roquin Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100655C
Description
Roquin Polyclonal specifically detects Roquin in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Roquin | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| KIAA2025RP5-1198E17.5, RING finger and C3H zinc finger protein 1, ring finger and CCCH-type domains 1, RING finger protein 198, RNF198ring finger and CCCH-type zinc finger domains 1, roquin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 149041 | |
| Store at 4C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RC3H1 | |
| The immunogen recognized by this antibody maps to a region between residues 500 and 550 of human Ring Finger and CCCH-type finder domains 1 using the numbering given in entry NP_742068.1 (GeneID 149041). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction