Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROR alpha/NR1F1 Antibody (784652), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8924H
Description
ROR alpha/NR1F1 Monoclonal antibody specifically detects ROR alpha/NR1F1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| ROR alpha/NR1F1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 784652 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| DKFZp686M2414, MGC119326, NR1F1MGC119329, nuclear receptor ROR-alpha, Nuclear receptor RZR-alpha, Nuclear receptor subfamily 1 group F member 1, RAR-related orphan receptor A, RAR-related orphan receptor alpha, retinoic acid receptor-related orphan receptor alpha, retinoid-related orphan receptor alpha, Retinoid-related orphan receptor-alpha, ROR1, ROR2, ROR3, RZR-ALPHA, RZRAROR-alpha, transcription factor RZR-alpha | |
| E. coli-derived recombinant human ROR alpha/NR1F1, Arg47-His163, Accession # P35398 | |
| RUO | |
| 6095 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction