Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROR beta Antibody (OTI1G1), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273930AF647
Description
ROR beta Monoclonal specifically detects ROR beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ROR beta | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RORB | |
Recombinant protein fragment corresponding to amino acids 1-260 of human ROR beta (NP_008845) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1G1 | |
Western Blot, Immunohistochemistry | |
NR1F2nuclear receptor ROR-beta, Nuclear receptor RZR-beta, Nuclear receptor subfamily 1 group F member 2, RAR-related orphan receptor B, RAR-related orphan receptor beta, retinoic acid-binding receptor beta, retinoid-related orphan receptor beta, Retinoid-related orphan receptor-beta, ROR-BETA, RZRBbA133M9.1, RZR-BETA | |
Mouse | |
Affinity Purified | |
GPCR | |
6096.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction