Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROR gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534163
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534163 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534163 Supplier Invitrogen™ Supplier No. PA534163
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Orangutan, Monkey, Dog, Rabbit (93%) Marmoset, Mouse, Rat, Bat, Bovine (86%).

ROR gamma is a 560 amino acid novel DNA-binding transcription factor belonging to the ROR/RZR orphan nuclear receptor subfamily. Reports suggest thatROR gammais a ligand activated transcription factor and may be essential for lymphoid organogenesis (lymph nodes and Peyer's patches) and may play an important regulatory role in controlling apoptosis during thymopoiesis. ROR gamma inhibits the expression of Fas ligand and IL2 and directs the differentiation program of proinflammatory IL-17+ T helper cells. ROR gamma shares 50 to 51% sequence identity with RORA and RORB and is 88% identical to mouse ROR gamma. It is highly expressed in skeletal muscle and at lower levels in various other tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROR gamma
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene RORC
Gene Accession No. P51449
Gene Alias IMD42; Nr1f3; Nuclear receptor ROR-gamma; nuclear receptor RZR-gamma; Nuclear receptor subfamily 1 group F member 3; RAR related orphan receptor C; RAR-related orphan nuclear receptor variant 2; RAR-related orphan receptor C; RAR-related orphan receptor C, isoform a; RAR-related orphan receptor gamma; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; retinoid-related orphan receptor-gamma; RORC; RORG; RORgamma; RZRG; RZR-GAMMA; Thor; Thymus orphan receptor; TOR
Gene Symbols RORC
Host Species Rabbit
Immunogen Synthetic 14 amino acid peptide from modulating domain of human ROR Gamma.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6097
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.