Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROR gamma/RORC/NR1F3 Antibody (4G419), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP224449PCP
Description
ROR gamma/RORC/NR1F3 Monoclonal specifically detects ROR gamma/RORC/NR1F3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
ROR gamma/RORC/NR1F3 | |
Monoclonal | |
PerCP | |
P51449 | |
RORC | |
A portion of amino acids 1-50 of human ROR gamma was used as the immunogen. | |
0.1 mL | |
GPCR | |
6097 | |
Human, Rat | |
IgG1 κ |
Flow Cytometry | |
4G419 | |
Flow Cytometry 1 μL/1 million cells | |
MGC129539, Nuclear receptor ROR gamma, Nuclear receptor ROR-gamma, Nuclear receptor RZR gamma, Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-Related Orphan Nuclear Receptor Variant 2, RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma, Retinoid-related orphan receptor-gamma, ROR gamma, RORG, RORGMGC129539, RZRG, RZRGRZR-GAMMA, TOR | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
The amino acid sequence used as immunogen is 100% homologous in Human, 86% in Mouse and Rat, 80% in Bovine, and 73% in Chicken. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction