Learn More
Invitrogen™ ROR gamma (t) Monoclonal Antibody (AFKJS-9), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Description
Description: The AFKJS-9 monoclonal antibody reacts with the mouse, human and rhesus monkey RORgamma(t) protein. RORgamma is a member of the retinoic acid-related orphan receptor (ROR) family, which also includes RORalpha and RORbeta. ROR family proteins are ligand-dependent transcription factors that play roles in multiple physiological processes. RORgamma is expressed in several tissues including liver, lung, muscle, heart and kidney. Furthermore, it was discovered that alternative transcription results in the expression of an isoform, RORgamma(t), which is expressed exclusively in cells of the lymphoid compartment, namely CD4+CD8+ double-positive thymocytes, Th17 cells of the periphery and lymphoid tissue inducer (LTi) cells of lymphoid organs. The RORgamma(t) isoform differs from RORgamma by three unique amino acids at its amino terminus. Therefore, the AFKJS-9 antibody will react with both the RORgamma and RORgamma(t) isoforms. Applications Reported: This AFKJS-9 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This AFKJS-9 antibody has been tested by intracellular staining followed by flow cytometric analysis of mouse thymocytes using the Foxp3/Transcription Factor Staining Buffer Set (Product No. 00-5523-00) and protocol.
Specifications
Specifications
| Antigen | ROR gamma (t) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | AFKJS-9 |
| Concentration | 0.2 mg/mL |
| Conjugate | PerCP-eFluor 710 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Rorc |
| Gene Accession No. | P51449, P51450 |
| Gene Alias | EGK_01272; IMD42; MGC129539; NR1F3; nuclear receptor ROR-gamma; Nuclear receptor RZR-gamma; nuclear receptor subfamily 1 group F member 3; RAR related orphan receptor C; RAR-related orphan nuclear receptor variant 2; RAR-related orphan receptor C; RAR-related orphan receptor C, isoform a; RAR-related orphan receptor gamma; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; retinoid-related orphan receptor-gamma; ROR Gamma; ROR gamma t; Rorc; Rorg; RORgamma; RORgt; RP11-98D18.11-001; RZRG; RZR-GAMMA; Thor; Thymus orphan receptor; TOR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.