Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROR gamma (t) Monoclonal Antibody (B2D), PE-Cyanine7, eBioscience™, Invitrogen™

Catalog No. 25698180
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-698-180 25 μg
25-698-182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-698-180 Supplier Invitrogen™ Supplier No. 25698180
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

The B2D monoclonal antibody reacts with the mouse ROR gamma (t) protein. ROR gamma is a member of the retinoic acid-related orphan receptor (ROR) family, which also includes ROR alpha and ROR beta. ROR family proteins are ligand-dependent transcription fa

ROR gamma, a NR1 Thyroid Hormone-Like receptor, has been shown to affect thymopoiesis, bone metabolism, T-cell apoptosis, and lymphoid organogenesis. ROR gamma has been shown to promote thymocyte survival by activating the expression of the antiapoptotic protein Bcl-x(L). ROR gamma is also required for the development of lymph nodes and Peyer's patches. It has been shown that ROR gamma(t), a thymus-specific isoform of ROR gamma from mouse, inhibits Fas ligand expression and cytokine secretion in immature thymocytes. ROR gamma binds as a monomer to response elements composed of a single core motif, GGTCA, preceded by a 6 bp AT-rich sequence. ROR gamma expression has been documented in mouse thymus, adipose, bone, skeletal muscle, liver, and kidney, and in human skeletal muscle. ESTs have been isolated from human tissue libraries, including cancerous head/neck, germ cell, and prostate, and normal colon, head/neck, lung, liver/spleen, and pancreas. ROR gammat t (RORgt) is an isoform of RORC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROR gamma (t)
Applications Flow Cytometry
Classification Monoclonal
Clone B2D
Concentration 0.2 mg/mL
Conjugate PE-Cyanine7
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Rorc
Gene Accession No. P51450
Gene Alias EGK_01272; IMD42; MGC129539; NR1F3; nuclear receptor ROR-gamma; Nuclear receptor RZR-gamma; nuclear receptor subfamily 1 group F member 3; RAR related orphan receptor C; RAR-related orphan nuclear receptor variant 2; RAR-related orphan receptor C; RAR-related orphan receptor C, isoform a; RAR-related orphan receptor gamma; retinoic acid-binding receptor gamma; retinoid-related orphan receptor gamma; retinoid-related orphan receptor-gamma; ROR Gamma; ROR gamma t; Rorc; Rorg; RORgamma; RORgt; RP11-98D18.11-001; RZRG; RZR-GAMMA; Thor; Thymus orphan receptor; TOR
Gene Symbols Rorc
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19885
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.