Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROR gamma t/RORC2/NR1F3 Antibody (1181A), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB61093G
Description
ROR gamma t/RORC2/NR1F3 Monoclonal antibody specifically detects ROR gamma t/RORC2/NR1F3 in Human samples. It is validated for Flow CytometrySpecifications
ROR gamma t/RORC2/NR1F3 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
6097 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
1181A | |
Intracellular Staining by Flow Cytometry | |
IMD42,NR1F3,Nuclear receptor ROR-gamma,RORG,RZRG,RZR-GAMMA,TOR | |
E.coli derived recombinant human ROR gamma T/RORC2/NR1F3, Met1-Gln100, Accession # P51449 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction