Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROR gamma t/RORC2/NR1F3 Antibody (1181A), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB61093MFV500
Description
ROR gamma t/RORC2/NR1F3 Monoclonal antibody specifically detects ROR gamma t/RORC2/NR1F3 in Human samples. It is validated for Flow CytometrySpecifications
| ROR gamma t/RORC2/NR1F3 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Flow Cytometry | |
| 1181A | |
| Intracellular Staining by Flow Cytometry | |
| IMD42;NR1F3;Nuclear receptor ROR-gamma;RORG;RZRG;RZR-GAMMA;TOR | |
| E.coli derived recombinant human ROR gamma T/RORC2/NR1F3, Met1-Gln100, Accession # P51449 | |
| RUO | |
| 6097 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction