Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROR1 Antibody (1011919), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB20001MFV610
Description
ROR1 Monoclonal antibody specifically detects ROR1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| ROR1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Protein Kinase | |
| 4919 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 1011919 | |
| Flow Cytometry, CyTOF-ready | |
| EC 2.7.10.1, MGC99659, neurotrophic tyrosine kinase receptor-related 1, Neurotrophic tyrosine kinase, receptor-related 1, NTRKR1dJ537F10.1, receptor tyrosine kinase-like orphan receptor 1, tyrosine-protein kinase transmembrane receptor ROR1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ROR1, Ala24-Glu403, Accession # Q01973 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction