Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR1 Mouse anti-Human, Brilliant Violet 421, Clone: 4A5, BD Optibuild™

Catalog No. BDB743012 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743012 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743012 Supplier BD Biosciences Supplier No. 743012
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 4A5 monoclonal antibody binds to the glycosylated type I membrane protein ROR1 (Receptor-tyrosine-kinase-like Orphan Receptor 1). The ROR1 and ROR2 related proteins are highly conserved evolutionarily and are primarily expressed during embryogenesis. ROR1 is not expressed on normal peripheral blood lymphocytes. ROR1 is similar to tropomyosin receptor kinase (Trk family) neurotropic receptors and shares a cysteine-rich domain with Frizzled receptors for Wnt-family signaling proteins, which are involved in the regulation of embryogenesis and carcinogenesis. Furthermore, there is mounting evidence that aberrant ROR1 expression contributes to human malignancy.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROR1
Classification Monoclonal
Clone 4A5
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Neurotrophic tyrosine kinase, receptor-related 1, NTRKR1
Host Species Mouse
Immunogen Human ROR1 cDNA
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.