Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR1 Mouse anti-Human, Brilliant Violet 786, Clone: 4A5, BD Optibuild™

Catalog No. BDB743017 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743017 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743017 Supplier BD Biosciences Supplier No. 743017
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 4A5 monoclonal antibody binds to the glycosylated type I membrane protein ROR1 (Receptor-tyrosine-kinase-like Orphan Receptor 1). The ROR1 and ROR2 related proteins are highly conserved evolutionarily and are primarily expressed during embryogenesis. ROR1 is not expressed on normal peripheral blood lymphocytes. ROR1 is similar to tropomyosin receptor kinase (Trk family) neurotropic receptors and shares a cysteine-rich domain with Frizzled receptors for Wnt-family signaling proteins, which are involved in the regulation of embryogenesis and carcinogenesis. Furthermore, there is mounting evidence that aberrant ROR1 expression contributes to human malignancy.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROR1
Classification Monoclonal
Clone 4A5
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Neurotrophic tyrosine kinase, receptor-related 1, NTRKR1
Host Species Mouse
Immunogen Human ROR1 cDNA
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.