Learn More
ROR1 Mouse anti-Human, Clone: OTI3D11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806826

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ROR1 is a receptor protein tyrosine kinase whose cellular role has not been determined. It is a type I membrane protein and belongs to the ROR subfamily of cell surface receptors. Studies of a similar protein in mouse suggest that this protein may interact with another receptor protein tyrosine kinase and may be involved in skeletal and cardiac development.Specifications
ROR1 | |
Monoclonal | |
Unconjugated | |
ROR1 | |
NTRKR1, dJ537F10.1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4919 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3D11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q01973 | |
ROR1 | |
Full length human recombit protein of human ROR1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.