Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR1 Mouse, anti-Human, R718, Clone: 4A5, BD OptiBuild™

Catalog No. BDB751907 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751907 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751907 Supplier BD Biosciences Supplier No. 751907
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 4A5 monoclonal antibody binds to the glycosylated type I membrane protein ROR1 (Receptor-tyrosine-kinase-like Orphan Receptor 1). The ROR1 and ROR2 related proteins are highly conserved evolutionarily and are primarily expressed during embryogenesis. ROR1 is not expressed on normal peripheral blood lymphocytes. ROR1 is similar to tropomyosin receptor kinase (Trk family) neurotropic receptors and shares a cysteine-rich domain with Frizzled receptors for Wnt-family signaling proteins, which are involved in the regulation of embryogenesis and carcinogenesis. Furthermore, there is mounting evidence that aberrant ROR1 expression contributes to human malignancy.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ROR1
Applications Flow Cytometry
Classification Monoclonal
Clone 4A5
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.