Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596241
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596241 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596241 Supplier Invitrogen™ Supplier No. PA596241
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549, 293T, HT-29, SKOV3; Cellular Location: Membrane, Single-pass type I membrane protein Immunogen sequence: ARGAAAQETE LSVSAELVPT SSWNISSELN KDSYLTLDEP MNNITTSLGQ TAELHCKVSG NPPPTIRWFK NDAPVVQEPR RLSFRSTIYG SRLRIRNLDT TDTGYFQCVA TNGKEVVSST GVLFVKFGPP PTASPGYSDE YEEDGFCQPY RGIACARFIG NRTVYMESLH MQGEIENQIT AAFTMIGTSS HLSDKCSQFA IPSLCHYAFP YCDETSSVPK PRDLCRDECE ILENVLCQTE YIFARSNPMI LMRLKLPNCE DLPQPESPEA ANCIRIGIPM ADPINKNHKC YNSTGVDYRG TVSVTKSGRQ CQPWNSQYPH THTFTALRFP ELNGGHSYCR NPGNQKEAPW CFTLDENFKS DLCDIPACDS KDSKEKNKME.

Receptor-tyrosine-kinase-like orphan receptor 1 (ROR1) is a tumor-associated, surface protein predominantly expressed during embryogenesis, where it is involved in organ morphogenesis, nervous system development, and neural progenitor cell maintenance and survival. Virtually absent from normal pediatric and adult tissues, with the exception of low-level expression in a subset of immature b cell precursors known as hematogones and adipocytes, ROR1 is notably overexpressed, and considered a survival factor, in a number of B lymphoid and epithelial malignancies: including chronic lymphocytic leukemia (CLL), mantle cell lymphoma (MCL), acute lymphoblastic leukemia (ALL), marginal zone lymphoma, lung adenocarcinoma. First identified during PCR-based cloning of a human neuroblastoma cell line in search of tyrosine kinases similar to tropomyosin-receptor-kinase (Trk) neurotropic receptors, ROR1, along with the related receptor tyrosine kinase (RTK) ROR2, was catalogued as an orphan receptor due to the fact its related ligand remained elusive. Wnt-5a has since been suggested as a candidate ligand for ROR1, and ROR1 has been implicated to function as a pseudokinase, promoting proliferation and resistance to apoptosis in cancer cells through interaction with Wnt-5a, and TCL1-co-activation of AKT. ROR1 is expressed as a glycoprotein containing extracellular immunoglobulin (Ig)-like, Frizzled, and Kringle domains, as well as an intracellular region containing a tyrosine kinase domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROR1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene ROR1
Gene Accession No. Q01973
Gene Alias 2810404D04Rik; dJ537F10.1; Inactive tyrosine-protein kinase transmembrane receptor ROR1; MGC99659; mRor1; neurotrophic tyrosine kinase, receptor related 1; neurotrophic tyrosine kinase, receptor-related 1; Ntrkr1; receptor tyrosine kinase like orphan receptor 1; receptor tyrosine kinase-like orphan receptor 1; RGD1559469; ROR1; T-ROR1; tyrosine-protein kinase transmembrane receptor ROR1
Gene Symbols ROR1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-403 of human ROR1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4919
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.