Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROR1 Polyclonal Antibody
ROR1 Polyclonal Antibody
Supplier: Thermo Scientific PA5105303
Description
ROR1 Polyclonal Antibody for Western Blot

Specifications
ROR1 | |
Polyclonal | |
Unconjugated | |
ROR1 | |
2810404D04Rik; dJ537F10.1; Inactive tyrosine-protein kinase transmembrane receptor ROR1; MGC99659; mRor1; neurotrophic tyrosine kinase, receptor related 1; neurotrophic tyrosine kinase, receptor-related 1; Ntrkr1; receptor tyrosine kinase like orphan receptor 1; receptor tyrosine kinase-like orphan receptor 1; RGD1559469; ROR1; T-ROR1; tyrosine-protein kinase transmembrane receptor ROR1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26563, 4919 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q01973, Q9Z139 | |
ROR1 | |
A synthesized peptide derived from human ROR1(Accession Q01973), corresponding to amino acid residues T760-I810. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction