Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROR2 Monoclonal Antibody (6F2D10)
GREENER_CHOICE

Catalog No. PIMA531899
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531899 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531899 Supplier Invitrogen™ Supplier No. MA531899
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31899 has been tested in indirect ELISA.

ROR2 (receptor tyrosine kinase-like orphan receptor 2) is a type I transmembrane protein that belongs to the ROR subfamily of cell surface receptors. It is thought to be involved in the early formation of the chondrocytes and cartilage and growth plate development. This Type I membrane protein is expressed at high levels during early embryonic development. The expression levels drop strongly around day 16 and there are only very low levels in adult tissues. Defects in ROR2 are a cause of brachydactyly type B1 (BDB1). BDB1 is an autosomal dominant skeletal disorder characterized by hypoplasia/aplasia of distal phalanges and nails. In BDB1 the middle phalanges are short but in addition the terminal phalanges are rudimentary or absent. Both fingers and toes are affected. The thumbs and big toes are usually deformed. Defects in ROR2 are a cause of recessive Robinow syndrome (RRS). RRS is an autosomal disorder characterized by skeletal dysplasia with generalized limb bone shortening, segmental defects of the spine, brachydactyly and a dysmorphic facial appearance. The protein contains 1 frizzled (FZ) domain, 1 immunoglobulin-like C2-type domain, and 1 kringle domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROR2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 6F2D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene ROR2
Gene Accession No. Q01974
Gene Alias BDB; BDB1; mRor2; neurotrophic tyrosine kinase receptor-related 2; neurotrophic tyrosine kinase, receptor related 2; neurotrophic tyrosine kinase, receptor-related 2; NTRKR2; receptor tyrosine kinase like orphan receptor 2; receptor tyrosine kinase-like orphan receptor 2; ROR2; Tyrosine-protein kinase transmembrane receptor ROR2
Gene Symbols ROR2
Host Species Mouse
Immunogen Purified recombinant fragment of human ROR2 (AA: 59-155) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4920
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.