Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROR2 Monoclonal Antibody (ROR2 2535-2835), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 34045100UG

This item is not returnable.
View return policy
Description
ROR2 Monoclonal antibody specifically detects ROR2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
ROR2 | |
Monoclonal | |
0.5-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q01974, Q9Z138 | |
Mouse | |
Protein A | |
RUO | |
26564, 4920 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
ROR2 2535-2835 | |
Unconjugated | |
ROR2 | |
BDB; BDB1; mRor2; neurotrophic tyrosine kinase receptor-related 2; neurotrophic tyrosine kinase, receptor related 2; neurotrophic tyrosine kinase, receptor-related 2; NTRKR2; receptor tyrosine kinase like orphan receptor 2; receptor tyrosine kinase-like orphan receptor 2; ROR2; Tyrosine-protein kinase transmembrane receptor ROR2 | |
Bases 2535-2835 of mouse ROR2 fused in-frame to MBP and expressed in E. coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction