Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RORA Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145075 Supplier Invitrogen™ Supplier No. PA5145075
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VQKHRMQQQQ RDHQQQPGEA EPLTPTYNIS ANGLTELHDD LSNYIDGHTP EGSKADSAVS SFYLDIQ Antigen sequence identity: Mouse 97%; Rat 97%.

RORA is a receptor for retinoic acid belonging to the NR1 subfamily of nuclear hormone receptors with a nuclear receptor DNA binding domain. RORA binds either as a monomer or as a homodimer to the retinoic acid response element and thus regulates gene expression and also controls cell function. Though the exact function of RORA is yet unknown, it interacts with NM23-2, a nucleoside diphosphate kinase involved in organogenesis and differentiation, NM23-1, the product of a tumor metastasis suppressor candidate gene, CDK7, NCOA3 and NCOA6 coactivators. It is found to express in mesenchymal stem cells derived from bone marrow and are known to act by direct modulation of a bone matrix component. It is widely expressed in most of the tissues. Chromosomal aberrations involving RORA may be a cause of acute promyelocytic leukemia (APL).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RORA
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RORA
Gene Accession No. P35398
Gene Alias 9530021D13Rik; nmf267; NR1F1; Nuclear receptor ROR-alpha; nuclear receptor RZR-alpha; Nuclear receptor subfamily 1 group F member 1; RAR related orphan receptor A; RAR-related orphan receptor A; RAR-related orphan receptor alpha; retinoic acid receptor-related orphan receptor alpha; retinoid-related orphan receptor alpha; retinoid-related orphan receptor-alpha; ROR Alpha; ROR1; ROR2; ROR3; Rora; Roralpha; ROR-alpha; RORalpha1; RZRA; RZR-ALPHA; sg; staggerer; thyroid hormone nuclear receptor alpha variant 4; tmgc26; transcription factor RZR-alpha
Gene Symbols RORA
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 160-226. Recombinant protein control fragment (Product #RP-110071).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6095
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.