Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RORA Polyclonal Antibody

Rabbit Polyclonal Antibody

$209.50 - $511.50

Specifications

Antigen RORA
Concentration 0.5 mg/mL
Content And Storage -20°C
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5119386
View Documents
Invitrogen™
PA5119386
200 μL
Each for $511.50
Only null left
Add to Cart
 
PIPA5121854
View Documents
Invitrogen™
PA5121854
20 μL
Each for $209.50
Only null left
Add to Cart
 
Description

Description

RORA Polyclonal Antibody for Western Blot, ICC/IF

RORA is a receptor for retinoic acid belonging to the NR1 subfamily of nuclear hormone receptors with a nuclear receptor DNA binding domain. RORA binds either as a monomer or as a homodimer to the retinoic acid response element and thus regulates gene expression and also controls cell function. Though the exact function of RORA is yet unknown, it interacts with NM23-2, a nucleoside diphosphate kinase involved in organogenesis and differentiation, NM23-1, the product of a tumor metastasis suppressor candidate gene, CDK7, NCOA3 and NCOA6 coactivators. It is found to express in mesenchymal stem cells derived from bone marrow and are known to act by direct modulation of a bone matrix component. It is widely expressed in most of the tissues. Chromosomal aberrations involving RORA may be a cause of acute promyelocytic leukemia (APL).
Specifications

Specifications

RORA
-20°C
Polyclonal
Liquid
IgG
Human
RORA
9530021D13Rik; nmf267; NR1F1; Nuclear receptor ROR-alpha; nuclear receptor RZR-alpha; Nuclear receptor subfamily 1 group F member 1; RAR related orphan receptor A; RAR-related orphan receptor A; RAR-related orphan receptor alpha; retinoic acid receptor-related orphan receptor alpha; retinoid-related orphan receptor alpha; retinoid-related orphan receptor-alpha; ROR Alpha; ROR1; ROR2; ROR3; Rora; Roralpha; ROR-alpha; RORalpha1; RZRA; RZR-ALPHA; sg; staggerer; thyroid hormone nuclear receptor alpha variant 4; tmgc26; transcription factor RZR-alpha
RORA
Primary
Antigen affinity chromatography
0.5 mg/mL
Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300
P35398
6095
Recombinant protein RAR Related Orphan Receptor Alpha. The antigen corresponds to amino acid range 164-401 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.