Learn More
RORB Mouse anti-Human, Clone: OTI4B7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806944

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Retinoid-related orphan receptor beta (ROR beta), a NR1 thyroid hormone receptor, affects the development and functioning of the central nervous system, including sensory input integration and circadian timing. Disruption of ROR beta in mice results in juvenile ataxia, retinal degeneration, circadian activity abnormalities, and delayed onset of male fertility. ROR beta binds to the monomeric response elements containing the sequence AnnTAGGTCA as a monomer. An alternatively spliced isoform, ROR beta2, has been isolated from rats, and the expression of ROR beta2 is confined to the pineal gland and retina. ROR beta2 shares common DNA- and putative ligand-binding domains with wild-type ROR beta but is characterized by a different amino-terminal domain. ROR beta expression has been documented in various regions of the mouse brain.Specifications
RORB | |
Monoclonal | |
Unconjugated | |
RORB | |
NR1F2, ROR-BETA, RZR-BETA, RZRB, bA133M9.1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6096 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4B7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q92753 | |
RORB | |
Human recombit protein fragment corresponding to amino acids 1-260 of human RORB produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.