Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RORB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534160
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534160 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534160 Supplier Invitrogen™ Supplier No. PA534160
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RORB Polyclonal Antibody for IHC (P)

Retinoid-related orphan receptor beta (ROR beta), a NR1 thyroid hormone receptor, affects the development and functioning of the central nervous system, including sensory input integration and circadian timing. Disruption of ROR beta in mice results in juvenile ataxia, retinal degeneration, circadian activity abnormalities, and delayed onset of male fertility. ROR beta binds to the monomeric response elements containing the sequence AnnTAGGTCA as a monomer. An alternatively spliced isoform, ROR beta2, has been isolated from rats, and the expression of ROR beta2 is confined to the pineal gland and retina. ROR beta2 shares common DNA- and putative ligand-binding domains with wild-type ROR beta but is characterized by a different amino-terminal domain. ROR beta expression has been documented in various regions of the mouse brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RORB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene RORB
Gene Accession No. P45446, Q8R1B8, Q92753
Gene Alias bA133M9.1; Nr1f2; Nuclear receptor ROR-beta; nuclear receptor RZR-beta; Nuclear receptor subfamily 1 group F member 2; RAR related orphan receptor B; RAR-related orphan receptor B; RAR-related orphan receptor beta; retinoic acid-binding receptor beta; retinoid-related orphan receptor b; retinoid-related orphan receptor beta; retinoid-related orphan receptor-beta; ror beta; Rorb; Rorbeta; ROR-BETA; rzrb; RZR-beta; sb:eu943; transcription factor; Unknown (protein for IMAGE:7173671)
Gene Symbols RORB
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from ligand-binding domain of human ROR Beta.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100049831, 100169935, 100345295, 100771338, 225998, 309288, 395980, 484164, 528382, 6096
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Sheep, Hamster, Horse, Chicken, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.