Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROS Antibody (4-6G), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250179AF594
Description
ROS Monoclonal specifically detects ROS in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ROS | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
ROS1 | |
Extracellular portion of ROS amino acid 1-285 fused to Fc, transiently expressed in 293 cells and purified using PtnA column chromatography. | |
0.1 mL | |
6098 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
4-6G | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
proto-oncogene tyrosine-protein kinase ROS, c-ros oncogene 1, receptor tyrosine kinase, c-ros-1, MCF3, ROS | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction