Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROS Antibody (7-2B), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250182PE
Description
ROS Monoclonal specifically detects ROS in Human samples. It is validated for Flow Cytometry.Specifications
ROS | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
ROS1 | |
Extracellular portion of ROS amino acid 1-285 fused to Fc, transiently expressed in 293 cells and purified using PtnA column chromatography. | |
0.1 mL | |
Protein Kinase | |
6098 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
7-2B | |
Flow Cytometry | |
proto-oncogene tyrosine-protein kinase ROS, c-ros oncogene 1 receptor tyrosine kinase, c-ros-1, MCF3, ROS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction