Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Roscovitine
GREENER_CHOICE

Catalog No. 133210
Change view
Click to view available options
Quantity:
10 mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-321-0 10 mg
13-325-0 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-321-0 Supplier Tocris Bioscience™ Supplier No. 1332/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective cyclin-dependent kinase inhibitor

Cyclin-dependent kinase (cdk) inhibitor. Displays sub-micromolar inhibition of cdk1, cdk2, cdk5, cdk7 and cdk9 (reported IC50 values range from 0.16 - 0.28 μM for cdk5 to 0.8 μM for cdk7). Selective for cdks over a wide range of related kinases including ERK1 and ERK2. Arrests L1210 cells in G1 phase. Inhibits phosphorylation of vimentin in vivo. Antimitotic.

Specifications

CAS 186692-46-6
Synonym roscovitine, seliciclib, r-roscovitine, rosco, 2-r-1-ethyl-2-hydroxyethylamino-6-benzylamino-9-isopropylpurine, unii-0es1c2kq94, insolution™ roscovitine, chembl14762, bmk1-e12
Quantity 10 mg
Molecular Formula C19H26N6O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.