Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Rose Bengal
SDP

Catalog No. 516850 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-685-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 51-685-0 Supplier Tocris Bioscience™ Supplier No. 5168/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent VGlut and VMAT inhibitor; cell permeable

Rose Bengal is a potent VGlut and vesicular monoamine transporter (VMAT) inhibitor (Ki values are 19 and 64 nM, respectively). Inhibits ATP dependent vesicular glutamate uptake (IC50 = 37 nM) and diminishes the amount of exocytotically released glutamate in synaptosomes. Decreases synaptic transmission in the dentate gyrus. Also stains exposed epithelium in the eye. Cell permeable.

Specifications

Purity >98%
Quantity 50 mg
Product Type Rose Bengal
Molecular Weight (g/mol) 1017.64
Inhibitors Rose Bengal

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.