Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Rostafuroxin
GREENER_CHOICE

Catalog No. 531510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-151-0 10 mg
53-155-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-151-0 Supplier Tocris Bioscience™ Supplier No. 5315/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Na+/K+ ATPase modulator; antihypertensive agent

Rostafuroxin is a Na+/K+ ATPase modulator. Inhibits the Normalizes renal Na+/K+ ATPase pump function in mutant adducin- and endogenous ouabain-dependent forms of hypertension. Reduces blood pressure in Milan hypertensive rats. Controls mania-like behaviors in Myshkin mice having a Na+/K+ ATPase α3 subunit mutation. Antagonizes the effects of Ouabain (Cat.No. 1076) on myogenic tone and Ca2+ signaling in rat hippocampal neurons.

Specifications

Chemical Name or Material 3β,5β,14β)-21,23-Epoxy-24-norchola-20,22-diene-3,14,17-triol
CAS 156722-18-8
Synonym PST 2238
Quantity 10 mg
Target Na+/K+ ATPase Modulators
Molecular Formula C23H34O4
Formula Weight 374.51
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.