Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Rosuvastatin calcium
GREENER_CHOICESDP

Catalog No. 634310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-431-0 10 mg
63-435-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-431-0 Supplier Tocris Bioscience™ Supplier No. 6343/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent HMG-CoA reductase inhibitor

Rosuvastatin calcium is a potent HMG-CoA reductase inhibitor (IC50 = 5.4 nM). Inhibits cholesterol synthesis in rat hepatocytes in vitro and in vivo. Reduces plasma LDL cholesterol levels. Enhances lentiviral transduction of natural killer cells. Orally bioavailable.

Specifications

Chemical Name or Material (3 R,5 S,6 E)-7-[4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidin-5-yl])-3,5-dihydroxyhept-6-enoic acid hemicalcium salt
CAS 147098-20-2
Assay Percent Range >99%
Quantity 10 mg
Target HMG-CoA Reductase Inhibitors
Molecular Formula C22H27FN3O6S.1⁄2Ca
Formula Weight Observed MW: 500.57
Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.