Learn More
BUCHI Rotavapor Cooling Water Valve
SDP
Catalog No. 01673004 Shop All BUCHI Products
Change view
Click to view available options
:
Rotavapor Cooling Water Valve; Type: Cooling Water Valve
Catalog No.
01-673-004 Rotavapor Cooling Water Valve; Type: Cooling Water Valve
1 options
Catalog No. 01-673-004 Supplier BUCHI Supplier No. 031356
Only null left
Add to Cart
Add to Cart
Automatically begin and end cooling media flow to your Rotavapor with this cooling water valve.
  • Valve to automatically turn on/off external cooling source
Specifications
Type Cooling Water Valve