Learn More
BUCHI Rotavapor R-300 Vertical Condenser Glass Assembly
SDP
Catalog No. p-7219081 Shop All BUCHI Products
Click to view available options
Material (Coating):
No Safety Coating
P+G Safety Coating
Catalog No. 05-405-067
Supplier: BUCHI 11062433
Only null left
Add to Cart
Add to Cart
Vertical Condenser Glass Assembly for Rotavapor R-300, 1500 cm2. Assembly features a large top inlet for easy cleaning, connection for vapor temperature sensor, and inner glass tubing/plate. Includes a 1 liter receiving flask, tubing, and a ball joint clamp.
  • Includes receiving flask (1 L), tubing, and a ball joint clamp
  • Does NOT include evaporating flask, vacuum seal, vapor duct, and condenser holder
  • For use with Rotavapor R-300, R-2XX
Specifications
Material Borosilicate Glass (3.3)
Type Vertical Condensor
For Use With (Application) Rotary Evaporator
For Use With (Equipment) Rotavap™ R-300, R-210, and R-215
Includes Receiving Flask, Tubing, and a Ball Joint Clamp
Material (Coating) P+G Safety Coating
Volume (Metric) 1 L
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.