Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rotavirus A MAb Mouse anti-All, Unconjugated, Clone: Clone 1, MilliporeSigma™

Catalog No. MABX84511MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABX84511MI 1 mg
MABX84110MI 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABX84511MI Supplier MilliporeSigma Supplier No. MABX84511KC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rotavirus A MAb Monoclonal antibody specifically detects Rotavirus A MAb in All samples. It is validated for Enzyme Immunoassay (ELISA), Immunodiffusion, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rotavirus A MAb
Applications ELISA, Immunodiffusion, Western Blot
Classification Monoclonal
Clone Clone 1
Conjugate Unconjugated
Gene Alias Anti-Rotavirus
Host Species Mouse
Immunogen Viral Lysate
Purification Method Affinity Purified
Quantity 1 mg
Research Discipline Infectious Diseases
Primary or Secondary Primary
Test Specificity Specific for intact virus of strains: Rhesus Rotavirus, human rotavirus WA strain, SA11, etc.
Target Species All species
Content And Storage 2 to 8°C. For long-term storage, freeze at -20°C.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.