Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rotavirus A VP6 Mouse anti-Virus, Clone: UK1, Abnova™

Catalog No. 89035556 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-556 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-556 Supplier Abnova Corporation Supplier No. MAB14158

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against Rotavirus A VP6.

  • High specificity
  • High reactivity

Specifications

Antigen Rotavirus A VP6
Applications Lateral Flow
Classification Monoclonal
Clone UK1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Rotavirus A VP6.
Dilution Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (< 15 mM sodium azide).
Gene Accession No. P04509
Gene Symbols RVA_s6gp1
Host Species Mouse
Immunogen Human Rotavirus, purified by caesium chloride (CsCl) gradient.
Purification Method Protein A affinity purification
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7011372
Target Species Virus
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.