Learn More
Invitrogen™ Rotavirus Monoclonal Antibody (3C10cc)
Description
MA1-46451 cross-reacts with monkey rotavirus (SA-11), porcine rotavirus (PP) and human rotaviruses (field strains).
Specifications
Specifications
| Antigen | Rotavirus |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 3C10cc |
| Concentration | 5.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene Alias | Reoviridae |
| Host Species | Mouse |
| Immunogen | Purified bovine rotavirus, cell culture adapted strain MR. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.