Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rotavirus Monoclonal Antibody (G7D)
GREENER_CHOICE

Catalog No. PIMA518251
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518251 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518251 Supplier Invitrogen™ Supplier No. MA518251
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with intact Rhesus rotavirus, WA (serotype 2) rotavirus, and Bovine rotavirus. Purity is >90%.

Rotavirus is a double-stranded RNA virus in the family Reoviridae. Five species of rotavirus (A-E) cause disease in animals with species A being the most common in human. Within rotavirus A there are different strains, called serotypes, based on the dual classification system of two proteins on the surface of the virus. Six viral proteins (VPs), or structural proteins, form the virion and six non-structural proteins (NSPs) are expressed in infected cells. Rotavirus is the most common cause of severe diarrhoea among infants and young children. Rotavirus gastroenteritis is a mild to severe disease characterised by vomiting, watery diarrhoea, and low-grade fever, but still leads to hundreds of thousands of deaths each year in developing countries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rotavirus
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone G7D
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Reoviridae
Host Species Mouse
Immunogen Rotavirus native protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.