Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rotavirus NCDV Polyclonal Antibody, HRP
GREENER_CHOICE

Catalog No. PIPA173015
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173015 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173015 Supplier Invitrogen™ Supplier No. PA173015
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The PA1-73015 antibody reacts with the ICP's and lat structural (viron) antigens of Nebraska Calf Diarrhea Virus. This antibody shows >90% cross reactivity with human rotaviruses. PA1-73015 has been successfully used in ELISA applications. The PA1-73015 antibody was raised against Nebraska Calf Diarrhea Virus. NOTE: Sodium azide is a potent inhibitor of peroxidase!.

Rotavirus is a double-stranded RNA virus in the family Reoviridae. Five species of rotavirus (A-E) cause disease in animals with species A being the most common in human. Within rotavirus A there are different strains, called serotypes, based on the dual classification system of two proteins on the surface of the virus. Six viral proteins (VPs), or structural proteins, form the virion and six non-structural proteins (NSPs) are expressed in infected cells. Rotavirus is the most common cause of severe diarrhoea among infants and young children. Rotavirus gastroenteritis is a mild to severe disease characterised by vomiting, watery diarrhoea, and low-grade fever, but still leads to hundreds of thousands of deaths each year in developing countries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rotavirus NCDV
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 10mg/mL BSA and 0.002% thimerosal
Gene Alias Reoviridae
Host Species Goat
Immunogen Nebraska Calf Diarrhea Virus.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.