Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rotavirus VP6 Mouse anti-Virus, Unconjugated, Clone: HM630, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation MAB21952
Description
Rotavirus VP6 Monoclonal antibody specifically detects Rotavirus VP6 in Viruses samples. It is validated for ELISA, LFIA.Specifications
Rotavirus VP6 | |
Monoclonal | |
Unconjugated | |
Recombinant Rotavirus VP6 | |
Primary | |
Store at 4°C | |
IgG1 |
ELISA, Lateral Flow | |
HM630 | |
Mouse | |
1 mg | |
Virus | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction