Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Rotigotine hydrochloride
GREENER_CHOICE

Catalog No. 389610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-961-0 10 mg
38-965-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-961-0 Supplier Tocris Bioscience™ Supplier No. 3896/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dopamine D2/D3 agonist

Dopamine D2 and D3 receptor agonist. Ki values are 13 and 0.71 nM for D2 and D3 respectively. Also has significant affinity for 5-HT1A and adrenergic α2B receptors. Exhibits antiparkinsonian acitivity.

Chemical Identifiers

CAS 125572-93-2
Molecular Formula C19H26ClNOS

Specifications

Quantity 10 mg
Formula Weight 351.93
Purity >99%
Molecular Weight (g/mol) 351.933
Chemical Name or Material Rotigotine hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.