Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rottlerin
GREENER_CHOICESDP

Catalog No. 161050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-105-0 50 mg
16-101-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-105-0 Supplier R&D Systems™ Supplier No. 1610/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Reported PKCδ inhibitor

Originally reported to inhibit PKC isoforms. Also reported to inhibit CAM kinase III. However, recently shown to inhibit a wide range of protein kinases, and most potently to inhibit PRAK and MAPKAP-K2 (IC50 values are 1.9 and 5 μM respectively). Also shown to act as a direct mitochondrial uncoupler. Thought to stimulate autophagy by targeting upstream mTORC1 control pathways.

Chemical Identifiers

CAS 82-08-6
Molecular Formula C30H28O8

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 516.55
Purity >98%
Molecular Weight (g/mol) 516.546
Chemical Name or Material Rottlerin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.