Learn More
Eppendorf™ Round Buckets for S-4x400 Rotor
Catalog No. 13864426 Shop All Eppendorf North America Products
Change view
Click to view available options
Quantity:
2 Pk.
4 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-864-426 4 Pk.
13-864-427 2 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-864-426 Supplier Eppendorf™ Supplier No. 5895182003
Add to Cart
Add to Cart
These Round Buckets for S-4x400 Rotor are suitable swing-buckets for the S-4x400 rotor that feature a round shape and a 400 mL capacity.
  • Suitable for S-4x400 rotor
  • Round shape
  • Swing-out bucket
  • Variable places depending on adapter used
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Rotor S-4x400
Capacity 400 mL
Quantity 4 Pk.