Learn More
Mopec™ Round Tissue Flotation Bath Lid
SDP
This Round Tissue Flotation Bath is designed with a deep center bowl, so slides can be fully submerged at a convenient angle. It features an anti-microbial coating to protect the system from bacterial and viral growth as well as a rapid heating system to reduce waiting time.
Supplier:  Mopec™ BK100L
Catalog No. 22-445-389
Only null left
Add to Cart
Description
Description
  • For use with the Mopec™ BK100 Round Tissue Flotation Bath
Specifications
Specifications
30°C to 70°C
Flotation Bath Lid
None
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.