Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Roxithromycin 95.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Type | Roxithromycin |
Physical Form | Crystalline Powder |
Melting Point | 120°C |
Color | White |
Quantity | 25 g |
Formula Weight | 837.06 |
Percent Purity | ≥95.0% (HPLC) |
Chemical Name or Material | Roxithromycin |
Safety and Handling
RTECSNumber | KF4990000 |
TSCA | No |
Recommended Storage | Refrigerator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction