Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RP-1 Antigen Mouse anti-Rat , APC, Clone: RP-1, BD Biosciences™

Catalog No. BDB566870 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566870 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566870 Supplier BD Biosciences Supplier No. 566870
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RP-1 Antigen Monoclonal antibody specifically detects RP-1 Antigen in Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen RP-1 Antigen
Applications Flow Cytometry
Classification Monoclonal
Clone RP-1
Conjugate APC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias RP1 Antigen; Rat Granulocytes; Rat Neutrophils
Host Species Mouse
Immunogen WKA/Hok rat peritoneal neutrophils
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.