Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RP 6306
GREENER_CHOICE

Catalog No. 803110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-311-0 10 mg
80-315-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 80-311-0 Supplier Tocris Bioscience™ Supplier No. 8031/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective PKMYT1 inhibitor

RP 6306 is a potent and selective PKMYT1 inhibitor (IC50 = 3.1 nM). In vivo, it inhibits CCNE1-amplified tumor cell growth in mouse xenograft models. RP 6306 is orally bioavailable.

Specifications

Chemical Name or Material (S)-2-Amino-1-(3-hydroxy-2,6-dimethylphenyl)-5,6-dimethylpyrrolo[2,3-b]pyridine-3-carboxamide
CAS 2719793-90-3
Quantity 10 mg
Target Cyclin-dependent Kinase Inhibitors
Molecular Formula C18H20N4O2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.