Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ RP 67580
GREENER_CHOICESDP

Catalog No. 163550 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-355-0 50 mg
16-351-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-355-0 Supplier R&D Systems™ Supplier No. 1635/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective NK1 antagonist

Potent and selective tachykinin NK1 receptor antagonist (Ki values are 2.9 nM and > 10 μM for rat NK1, and rat NK2 and NK3 receptors respectively). Displays higher affinity at rat and mouse than human receptors. Antinociceptive in vivo, possibly partly via inhibition of calcium channels.

Chemical Identifiers

CAS 135911-02-3
Molecular Formula C29H30N2O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 438.57
Purity >97%
Molecular Weight (g/mol) 438.571
Chemical Name or Material RP 67580
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.